4-Bromo-2-pyridone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine

072157-1G 072157-5G 072157-25G
102523-394EA 106.15 USD
102523-394 102523-396 102833-524
4-Bromo-2-pyridone ≥95%
4-Bromo-2-pyridone

Hazard: Irritant

Formula: C₅H₄BrNO
Melting Pt: 201-204°C
MDL Number: MFCD00234041
CAS Number: 36953-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR