4-Bromo-1H-indole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

072156-1G 072156-5G
102523-390EA 238.46 USD
102523-390 102523-392
4-Bromo-1H-indole-2-carboxylic acid ≥95%
4-Bromo-1H-indole-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆BrNO₂
MDL Number: MFCD02664468
CAS Number: 16732-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR