4-Amino-3-hydroxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-4-aminopyridine, 4-Aminopyridin-3-ol

072155-250MG 072155-1G 072155-5G
102523-382EA 355.38 USD
102523-382 102523-384 102523-386
4-Amino-3-hydroxypyridine ≥95%
4-Amino-3-hydroxypyridine

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD01941262
CAS Number: 52334-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR