4-Amino-3-bromoquinoline ≥95%

Supplier: Matrix Scientific

072154-1G 072154-5G
102523-378EA 640 USD
102523-378 102523-380
4-Amino-3-bromoquinoline ≥95%
4-Amino-3-bromoquinoline

Hazard: Irritant

Formula: C₉H₇BrN₂
MDL Number: MFCD00234502
CAS Number: 36825-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR