4-Amino-2-fluoropyridine ≥95%

Supplier: Matrix Scientific

072152-1G 072152-5G
102523-368EA 33.85 USD
102523-368 102523-370
4-Amino-2-fluoropyridine ≥95%
4-Amino-2-fluoropyridine

Hazard: Irritant

Formula: C₅H₅FN₂
MW: 112.11 g/mol
MDL Number: MFCD03095284
CAS Number: 18614-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR