4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride

072151-1G 072151-5G
102523-362EA 330.77 USD
102523-362 102523-364
4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride ≥95%
4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD07772867
CAS Number: 106261-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR