4,7-Diazaindole ≥95%

Supplier: Matrix Scientific

Synonyms: 5H-Pyrrolo[2,3-b]pyrazine

072150-250MG 072150-1G 072150-5G
102523-356EA 769.23 USD
102523-356 102523-358 102523-360
4,7-Diazaindole ≥95%
4,7-Diazaindole

Hazard: Irritant

Formula: C₆H₅N₃
MW: 119.124 g/mol
MDL Number: MFCD09834819
CAS Number: 42430-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR