4-(4-Methylpiperazin-1-yl)benzylamine ≥95%

Supplier: Matrix Scientific

072145-1G 072145-5G
102523-332EA 355.38 USD
102523-332 102523-334
4-(4-Methylpiperazin-1-yl)benzylamine ≥95%
4-(4-Methylpiperazin-1-yl)benzylamine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD03086117
CAS Number: 216144-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR