4-(3,4-Dichlorophenyl)-1-tetralone ≥95%

Supplier: Matrix Scientific

072144-5G 072144-10G 072144-100G
102523-326EA 61.54 USD
102523-326 102523-328 102523-330
4-(3,4-Dichlorophenyl)-1-tetralone ≥95%
4-(3,4-Dichlorophenyl)-1-tetralone

Hazard: Irritant

Formula: C₁₆H₁₂Cl₂O
MDL Number: MFCD02093088
CAS Number: 79560-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR