3-O-ethyl-L-ascorbic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-5-((S)-1,2-Dihydroxyethyl)-4-ethoxy-3-hydroxyfuran-2(5H)-one

072143-1G 072143-5G 072143-10G
102523-320EA 178.46 USD
102523-320 102523-322 102523-324
3-O-ethyl-L-ascorbic acid ≥95%
3-O-ethyl-L-ascorbic acid

Hazard: Irritant

Formula: C₈H₁₂O₆
MW: 204.18 g/mol
MDL Number: MFCD09261382
CAS Number: 86404-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR