3-Methoxy-4-methyl-2-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

072142-250MG 072142-1G
102523-316EA 498.46 USD
102523-316 102523-318
3-Methoxy-4-methyl-2-nitrobenzoic acid ≥95%
3-Methoxy-4-methyl-2-nitrobenzoic acid

Hazard: Irritant

Formula: C₉H₉NO₅
MDL Number: MFCD09834816
CAS Number: 57281-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR