3-Maleimidopropionic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid, 3-Maleimido-propionic acid, N-(2-Carboxyethyl)maleimide, MPS-Acid, N-Maleoyl-β-alanine

072141-1G 072141-5G
102523-310EA 60 USD
102523-310 102523-312
3-Maleimidopropionic acid ≥95%
3-Maleimidopropionic acid

Hazard: Irritant/Moisture Sensitive/Stored under Argon

Formula: C₇H₇NO₄
MW: 169.14 g/mol
Melting Pt: 108…112 °C
MDL Number: MFCD00043030
CAS Number: 7423-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR