Gemcitabine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

072139-1G 072139-5G 072139-10G
102523-298EA 792.31 USD
102523-298 102523-300 102523-302
Gemcitabine ≥95%
Gemcitabine

Hazard: Irritant/Store in Freezer

Formula: C₉H₁₁F₂N₃O₄
MW: 263.2 g/mol
MDL Number: MFCD00869720
CAS Number: 95058-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR