3-Chloroisonicotinonitrile ≥95%

Supplier: Matrix Scientific

072138-1G 072138-5G
102523-292EA 49.23 USD
102523-292 102523-294
3-Chloroisonicotinonitrile ≥95%
3-Chloroisonicotinonitrile

Hazard: Toxic

Formula: C₆H₃ClN₂
MW: 138.56 g/mol
Melting Pt: 71…74 °C
MDL Number: MFCD05663706
CAS Number: 68325-15-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR