3-Bromoisoxazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

072136-250MG 072136-1G
102523-284EA 235.38 USD
102523-284 102523-286
3-Bromoisoxazole-5-carbaldehyde ≥95%
3-Bromoisoxazole-5-carbaldehyde

Hazard: Irritant

Formula: C4H2BrNO2
MW: 175.9700 g/mol
MDL Number: MFCD09834813
CAS Number: 220780-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR