3-Bromo-4-nitropyridine ≥95%

Supplier: Matrix Scientific

072135-1G 072135-5G
102523-280EA 90.77 USD
102523-280 102523-282
3-Bromo-4-nitropyridine ≥95%
3-Bromo-4-nitropyridine

Hazard: Irritant

Formula: C5H3BrN2O2
MDL Number: MFCD03085767
CAS Number: 89364-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR