3-Bromoisonicotinonitrile ≥95%
Supplier: Matrix Scientific
072134-1G
072134-5G
102523-274EA
90.77
USD
102523-274
102523-276
3-Bromoisonicotinonitrile ≥95%
3-Bromoisonicotinonitrile
Hazard: Irritant
Formula:
C₆H₃BrN₂ MW: 183.01 g/mol Melting Pt: 92…94 °C |
MDL Number:
MFCD00234143 CAS Number: 13958-98-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...