3-Bromo-o-anisaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-methoxybenzaldehyde

072133-1G 072133-5G
102523-266EA 176.92 USD
102523-266 102523-268
3-Bromo-o-anisaldehyde ≥95%
3-Bromo-o-anisaldehyde

Hazard: Irritant

Formula: C₈H₇BrO₂
MW: 215.0400 g/mol
MDL Number: MFCD09834811
CAS Number: 88275-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR