3-Amino-5-hydroxypyridine dihydrochloride ≥95%
Supplier: Matrix Scientific
072131-250MG
072131-1G
072131-5G
102523-254EA
569.23
USD
102523-254
102523-256
102523-258
3-Amino-5-hydroxypyridine dihydrochloride ≥95%
3-Amino-5-hydroxypyridine dihydrochloride
Hazard: Irritant
Formula:
C₅H₈Cl₂N₂O MW: 183.04 g/mol |
MDL Number:
MFCD09702444 CAS Number: 1186663-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...