3-Amino-5-chloropyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Chloropyridin-3-amine
072130-1G
072130-5G
102523-248EA
44.62
USD
102523-248
102523-250
3-Amino-5-chloropyridine ≥95%
3-Amino-5-chloropyridine
Hazard: Irritant
Formula:
C₅H₅ClN₂ MW: 128.56 g/mol |
MDL Number:
MFCD03701386 CAS Number: 22353-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...