4-Ethyl-1H-pyrazol-3-amine oxalate ≥95%

Supplier: Matrix Scientific

072129-1G 072129-5G
102523-244EA 427.69 USD
102523-244 102523-246
4-Ethyl-1H-pyrazol-3-amine oxalate ≥95%
4-Ethyl-1H-pyrazol-3-amine oxalate

Hazard: Irritant

Formula: C₇H₁₁N₃O₄
MDL Number: MFCD11114053
CAS Number: 1010800-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR