3,5-Dibromo-N,N-dimethylpyrazinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dibromo-N,N-dimethylpyrazin-2-amine

072128-1G 072128-5G
102523-240EA 533.85 USD
102523-240 102523-242
3,5-Dibromo-N,N-dimethylpyrazinamine ≥95%
3,5-Dibromo-N,N-dimethylpyrazinamine

Hazard: Irritant

Formula: C₆H₇Br₂N₃
MW: 280.96 g/mol
MDL Number: MFCD09842809
CAS Number: 84539-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR