BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester) ≥95%

Supplier: Matrix Scientific

Synonyms: N-(3-Maleimidopropionyloxy)succinimide, 2,5-Dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoate, N-Succinimidyl 3-Maleimidopropionat, 3-Maleimidopropionic Acid N-Succinimidyl Ester, N-Succinimidyl 3-maleimidopropionate, 3-(Maleimido)propionic acid N-hydroxysuccinimide ester, N-β-maleimidopropyl-oxysuccinimide ester, BMPS

072126-1G 072126-5G
102523-230EA 58.46 USD
102523-230 102523-232
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester) ≥95%
BMPS (3-Maleimidopropionic acid N-hydroxysuccinimide ester)

Hazard: Irritant/Moisture Sensitive/Refrigerate

Formula: C₁₁H₁₀N₂O₆
MW: 266.21 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD00043141
CAS Number: 55750-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR