3-(4-Methylpiperazin-1-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

072125-1G 072125-5G
102523-226EA 355.38 USD
102523-226 102523-228
3-(4-Methylpiperazin-1-yl)benzoic acid ≥95%
3-(4-Methylpiperazin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₂
MDL Number: MFCD06742238
CAS Number: 215309-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR