3-(4-Methylpiperazin-1-yl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3-Aminophenyl)-4-methylpiperazine, 3-(4-Methyl-1-piperazinyl)aniline

072124-1G 072124-5G 072124-10G
102523-220EA 15.38 USD
102523-220 102523-222 102523-224
3-(4-Methylpiperazin-1-yl)aniline ≥95%
3-(4-Methylpiperazin-1-yl)aniline

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MW: 191.28 g/mol
MDL Number: MFCD08435857
CAS Number: 148546-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR