2-Thiophenesulfonamide ≥95%

Supplier: Matrix Scientific

Synonyms: Thiophene-2-sulphonamide

072123-1G 072123-5G 072123-10G
102523-214EA 143.08 USD
102523-214 102523-216 102523-218
2-Thiophenesulfonamide ≥95%
2-Thiophenesulfonamide

Hazard: Irritant

Formula: C₄H₅NO₂S₂
MW: 163.21 g/mol
MDL Number: MFCD00185853
CAS Number: 6339-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR