2-Oxo-1-piperazineacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Oxopiperazin-1-yl)acetic acid

072122-1G 072122-5G
102523-210EA 184.62 USD
102523-210 102523-212
2-Oxo-1-piperazineacetic acid ≥95%
2-Oxo-1-piperazineacetic acid

Hazard: Irritant

Formula: C₆H₁₀N₂O₃
MW: 158.16 g/mol
MDL Number: MFCD09838956
CAS Number: 24860-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR