2-Methylsulfanylpyrimidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

072120-1G 072120-5G
102523-200EA 278.46 USD
102523-200 102523-202
2-Methylsulfanylpyrimidine-4-carboxylic acid ≥95%
2-Methylsulfanylpyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₆N₂O₂S
MDL Number: MFCD09834808
CAS Number: 1126-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR