2-Methylsulfanylpyrimidine-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Methylthio)pyrimidine-4-carbaldehyde

072119-250MG 072119-1G 072119-5G
102523-194EA 355.38 USD
102523-194 102523-196 102523-198
2-Methylsulfanylpyrimidine-4-carbaldehyde ≥95%
2-Methylsulfanylpyrimidine-4-carbaldehyde

Hazard: Irritant

Formula: C₆H₆N₂OS
MW: 154.19 g/mol
MDL Number: MFCD02683211
CAS Number: 1074-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR