2-Methyl-5-bromopyrimidine-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

072116-250MG 072116-1G 072116-5G
102523-178EA 427.69 USD
102523-178 102523-180 102523-182
2-Methyl-5-bromopyrimidine-4-carboxylic acid ≥95%
2-Methyl-5-bromopyrimidine-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₅BrN₂O₂
MDL Number: MFCD00466678
CAS Number: 100707-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR