2-Chloro-4-fluorobenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-fluorobenzo[d]thiazole

072112-1G 072112-5G
102523-156EA 533.85 USD
102523-156 102523-158
2-Chloro-4-fluorobenzothiazole ≥95%
2-Chloro-4-fluorobenzothiazole

Hazard: Irritant

Formula: C₇H₃ClFNS
MW: 187.62 g/mol
MDL Number: MFCD04448820
CAS Number: 182344-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR