2-Bromobenzo[d]thiazole-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

072110-1G 072110-5G
102523-146EA 710.77 USD
102523-146 102523-148
2-Bromobenzo[d]thiazole-6-carboxylic acid ≥95%
2-Bromobenzo[d]thiazole-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₄BrNO₂S
MW: 258.09 g/mol
MDL Number: MFCD09266217
CAS Number: 22514-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR