2-Bromo-4-hydroxypyridine ≥95%

Supplier: Matrix Scientific

072106-1G 072106-5G 072106-10G
102523-122EA 107.69 USD
102523-122 102523-124 102523-126
2-Bromo-4-hydroxypyridine ≥95%
2-Bromo-4-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄BrNO
MDL Number: MFCD00234040
CAS Number: 36953-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR