2-Bromonicotinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3-cyanopyridine

072104-1G 072104-5G
102523-112EA 36.92 USD
102523-112 102523-114
2-Bromonicotinonitrile ≥95%
2-Bromonicotinonitrile

Hazard: Irritant

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
MDL Number: MFCD00234140
CAS Number: 20577-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR