2-Amino-5-bromo-4,6-dimethylpyrimidine ≥95%

Supplier: Matrix Scientific

072102-1G 072102-5G 072102-10G
102523-102EA 284.62 USD
102523-102 102523-104 102523-106
2-Amino-5-bromo-4,6-dimethylpyrimidine ≥95%
2-Amino-5-bromo-4,6-dimethylpyrimidine

Hazard: Irritant

Formula: C₆H₈BrN₃
MDL Number: MFCD00160498
CAS Number: 4214-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR