2-Amino-4-hydroxypyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminopyrimidin-4-ol

072101-10G 072101-100G
102523-098EA 533.85 USD
102523-098 102523-100
2-Amino-4-hydroxypyrimidine ≥95%
2-Amino-4-hydroxypyrimidine

Hazard: Irritant

Formula: C₄H₅N₃O
MW: 111.1 g/mol
MDL Number: MFCD00023256
CAS Number: 100643-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR