2-Amino-3-methylpyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methylpyrazin-2-amine

072098-1G 072098-5G
102523-084EA 136.92 USD
102523-084 102523-086
2-Amino-3-methylpyrazine ≥95%
2-Amino-3-methylpyrazine

Hazard: Irritant

Formula: C₅H₇N₃
MW: 109.132 g/mol
MDL Number: MFCD08705770
CAS Number: 19838-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR