2-Amino-3-hydroxypyrazine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Aminopyrazin-2(1H)-one

072097-1G 072097-5G
102523-080EA 710.77 USD
102523-080 102523-082
2-Amino-3-hydroxypyrazine ≥95%
2-Amino-3-hydroxypyrazine

Hazard: Irritant

Formula: C₄H₅N₃O
MW: 111.1 g/mol
MDL Number: MFCD01646052
CAS Number: 43029-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR