2-Amino-3-chloro-5-methylpyrazine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Chloro-5-methylpyrazin-2-amine
072096-250MG
072096-1G
072096-5G
102523-074EA
640
USD
102523-074
102523-076
102523-078
2-Amino-3-chloro-5-methylpyrazine ≥95%
2-Amino-3-chloro-5-methylpyrazine
Hazard: Irritant
Formula:
C₅H₆ClN₃ MW: 143.5700 g/mol |
MDL Number:
MFCD09834805 CAS Number: 89182-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...