Piperidine-2,4-dione ≥95%
Supplier: Matrix Scientific
Synonyms:
2,4-Piperidinedione, 2,4-Dioxopiperidine
072091-1G
072091-5G
072091-25G
102523-044EA
36.92
USD
102523-044
102523-046
102551-790
Piperidine-2,4-dione ≥95%
Piperidine-2,4-dione
Hazard: Irritant/Stench
Formula:
C₅H₇NO₂ MW: 113.1146 g/mol Melting Pt: 98-103°C |
MDL Number:
MFCD08704814 CAS Number: 50607-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...