1-Phenyl-1,2,3,4-tetrahydroisoquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2,3,4-Tetrahydro-1-phenylisoquinoline

072086-1G 072086-5G
102523-018EA 193.85 USD
102523-018 102523-020
1-Phenyl-1,2,3,4-tetrahydroisoquinoline ≥95%
1-Phenyl-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₁₅H₁₅N
MW: 209.29 g/mol
MDL Number: MFCD02179241
CAS Number: 22990-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR