1-Boc-5-aminoindole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-1H-indole, N1-BOC protected, 1-Boc-5-Aminoindole

072083-1G 072083-5G 072083-10G
102523-002EA 250.77 USD
102523-002 102523-004 102523-006
1-Boc-5-aminoindole ≥95%
1-Boc-5-aminoindole

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₂
MW: 232.28
MDL Number: MFCD04114746
CAS Number: 166104-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR