1-Benzofuran-2-carbonitrile ≥95%

Supplier: Matrix Scientific

072080-1G 072080-5G
102522-988EA 427.69 USD
102522-988 102522-990
1-Benzofuran-2-carbonitrile ≥95%
1-Benzofuran-2-carbonitrile

Hazard: Irritant

Formula: C₉H₅NO
MDL Number: MFCD01659779
CAS Number: 41717-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR