7-Nitro-1,2,3,4-tetrahydroquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2,3,4-Tetrahydro-7-nitroquinoline

072079-1G 072079-5G 072079-10G
102522-982EA 161.54 USD
102522-982 102522-984 102522-986
7-Nitro-1,2,3,4-tetrahydroquinoline ≥95%
7-Nitro-1,2,3,4-tetrahydroquinoline

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
MDL Number: MFCD00496654
CAS Number: 30450-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR