(S)-Pyrrolidine-2-carbonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

072078-1G 072078-5G
102522-978EA 227.69 USD
102522-978 102522-980
(S)-Pyrrolidine-2-carbonitrile hydrochloride ≥95%
(S)-Pyrrolidine-2-carbonitrile hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₂
MDL Number: MFCD02683060
CAS Number: 65732-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR