(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-N-Despropyl Pramipexole, Pramipexole Related Compound A

072077-1G 072077-5G
102522-972EA 38.46 USD
102522-972 102522-974
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole ≥95%
(S)-(-)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole

Hazard: Irritant

Formula: C₇H₁₁N₃S
MW: 169.25 g/mol
MDL Number: MFCD07368003
CAS Number: 106092-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR