(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline

072075-250MG 072075-1G 072075-5G
102522-962EA 355.38 USD
102522-962 102522-964 102522-966
(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline ≥95%
(1S)-1-Phenyl-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₁₅H₁₅N
MW: 209.29 g/mol
MDL Number: MFCD08692036
CAS Number: 118864-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR