5-Amino-2-nitropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Nitropyridin-3-amine

072074-250MG 072074-1G 072074-5G
102522-956EA 498.46 USD
102522-956 102522-958 102522-960
5-Amino-2-nitropyridine ≥95%
5-Amino-2-nitropyridine

Hazard: Irritant

Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
MDL Number: MFCD01646117
CAS Number: 14916-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR