3-Hydroxypyridine-4-carboxaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxyisonicotinaldehyde

072073-250MG 072073-1G 072073-5G
102522-950EA 256.92 USD
102522-950 102522-952 102522-954
3-Hydroxypyridine-4-carboxaldehyde ≥95%
3-Hydroxypyridine-4-carboxaldehyde

Hazard: Irritant

Formula: C6H5NO2
MW: 123.1094 g/mol
MDL Number: MFCD01318565
CAS Number: 1849-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR