3-Fluoro-5-hydroxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-5-pyridinol, 5-Fluoropyridin-3-ol

072072-1G 072072-5G
102522-946EA 87.69 USD
102522-946 102522-948
3-Fluoro-5-hydroxypyridine ≥95%
3-Fluoro-5-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄FNO
MDL Number: MFCD03095199
CAS Number: 209328-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR